ENA371463596 Enamine

ethyl 2-(4-methoxyphenyl)-2-methylpropanoate

  • MDL number MFCD16756810
  • Cas Number : 2901-19-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109745
Availability details for
Quantity
  • Availability
  • Add to Cart