ENA371463604 Enamine

3-ethyl-3-methylmorpholine

  • MDL number MFCD09722899
  • Cas Number : 938326-10-4
  • Molecular Weight : 129.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-ethyl-3-methylmorpholine
Inchi Code 1S/C7H15NO/c1-3-7(2)6-9-5-4-8-7/h8H,3-6H2,1-2H3
Inchi Key XESSAGYMZVFWNO-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-109788

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-109788
pictograms GHS02,GHS05,GHS07
hstatements H226,H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart