ENA371463609 Enamine

4-methyl-3-(4-methyl-1,3-oxazol-2-yl)aniline

  • MDL number MFCD20306730
  • Cas Number : 1339691-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109804
Availability details for
Quantity
  • Availability
  • Add to Cart