ENA371463612 Enamine

3-methoxy-2-(trifluoromethyl)pyridine

  • MDL number MFCD13185702
  • Cas Number : 1184172-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104857
Availability details for
Quantity
  • Availability
  • Add to Cart