ENA371463620 Enamine

1-(3-fluoropyridin-2-yl)-1H-pyrazol-4-amine

  • MDL number MFCD16859677
  • Cas Number : 1339916-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109822
Availability details for
Quantity
  • Availability
  • Add to Cart