ENA371463632 Enamine

2-(1,2,3,4-tetrahydroisoquinolin-2-yl)pyridin-4-amine

  • MDL number MFCD16507769
  • Cas Number : 524718-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109841
Availability details for
Quantity
  • Availability
  • Add to Cart