ENA371463638 Enamine

2-bromo-1-(4-chlorophenyl)butan-1-one

  • MDL number MFCD12031566
  • Cas Number : 1011-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109851

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109851
Availability details for
Quantity
  • Availability
  • Add to Cart