ENA371463640 Enamine

2-(trifluoromethyl)-1,3-benzoxazol-4-amine

  • MDL number MFCD11857163
  • Cas Number : 573759-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109853

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109853
Availability details for
Quantity
  • Availability
  • Add to Cart