ENA371463647 Enamine

N-ethyl-N-(2-hydroxyethyl)pentanamide

  • MDL number MFCD10048253
  • Cas Number : 1011571-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109869
Availability details for
Quantity
  • Availability
  • Add to Cart