ENA371463654 Enamine

1,2,5,6,7,8-hexahydroquinoline-2,5-dione

  • MDL number MFCD00766821
  • Cas Number : 15450-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 289-291
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109919
Availability details for
Quantity
  • Availability
  • Add to Cart