ENA371463661 Enamine

methyl 2-amino-5-methyl-3-nitrobenzoate

  • MDL number MFCD20037605
  • Cas Number : 1248541-72-1
  • Molecular Weight : 210.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-108338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108338
Availability details for
Quantity
  • Availability
  • Add to Cart