ENA371463664 Enamine

5-fluoro-3-methoxy-1H-indole

  • MDL number MFCD20684049
  • Cas Number : 169674-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109934
Availability details for
Quantity
  • Availability
  • Add to Cart