ENA371463669 Enamine

9-azabicyclo[6.2.0]decane

  • MDL number MFCD21758456
  • Cas Number : 7489-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108354
Availability details for
Quantity
  • Availability
  • Add to Cart