ENA371463670 Enamine

2-hydroxy-2-(oxolan-3-yl)acetonitrile

  • MDL number MFCD12169349
  • Cas Number : 1155915-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109944
Availability details for
Quantity
  • Availability
  • Add to Cart