ENA371463673 Enamine

2-(3,5-dichlorophenyl)-2-(diethylamino)acetic acid

  • MDL number MFCD12517066
  • Cas Number : 1218081-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109947
Availability details for
Quantity
  • Availability
  • Add to Cart