ENA371463675 Enamine

3-(isocyanatomethyl)-1,3-diazinane-2,4-dione

  • MDL number MFCD11934276
  • Cas Number : 861359-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-109953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109953
Availability details for
Quantity
  • Availability
  • Add to Cart