ENA371463686 Enamine

1,5-diphenylpentan-1-amine

  • MDL number MFCD16183992
  • Cas Number : 21493-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109996

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-109996
Availability details for
Quantity
  • Availability
  • Add to Cart