ENA371463690 Enamine

4-[2-(1-aminoethyl)-1,3-thiazol-4-yl]benzonitrile

  • MDL number MFCD14628785
  • Cas Number : 1249162-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110001

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110001
Availability details for
Quantity
  • Availability
  • Add to Cart