ENA371463692 Enamine

1-[(benzylamino)methyl]cyclopentan-1-ol

  • MDL number MFCD17219385
  • Cas Number : 1310294-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108440

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108440
Availability details for
Quantity
  • Availability
  • Add to Cart