ENA371463693 Enamine

2-[1-(aminomethyl)cyclobutyl]acetic acid hydrochloride

  • MDL number MFCD20645823
  • Cas Number : 223425-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110004

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110004
Availability details for
Quantity
  • Availability
  • Add to Cart