ENA371463696 Enamine

1-[4-(3-chloro-4-fluorophenyl)-1,3-thiazol-2-yl]ethan-1-amine

  • MDL number MFCD14628790
  • Cas Number : 1248704-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110007

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110007
Availability details for
Quantity
  • Availability
  • Add to Cart