ENA371463708 Enamine

1-(propan-2-yl)-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD00957561
  • Cas Number : 77796-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110022

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110022
Availability details for
Quantity
  • Availability
  • Add to Cart