ENA371463718 Enamine

2,2,2-trifluoro-1-(piperidin-4-yl)ethan-1-ol hydrochloride

  • MDL number MFCD18800779
  • Cas Number : 184042-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110084
Availability details for
Quantity
  • Availability
  • Add to Cart