ENA371463720 Enamine

3-bromo-1-chloroprop-1-ene

  • MDL number MFCD01742789
  • Cas Number : 3737-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110086

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110086
Availability details for
Quantity
  • Availability
  • Add to Cart