ENA371463723 Enamine

2-fluoro-4-nitrobenzene-1-sulfonamide

  • MDL number MFCD16040222
  • Cas Number : 1204574-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-147
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110099
Availability details for
Quantity
  • Availability
  • Add to Cart