ENA371463728 Enamine

5-fluoro-3-methyl-1-benzofuran

  • MDL number MFCD13191783
  • Cas Number : 33118-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100325

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100325
Availability details for
Quantity
  • Availability
  • Add to Cart