ENA371463729 Enamine

2,2-diethyl-3-methoxycyclobutan-1-amine

  • MDL number MFCD21757669
  • Cas Number : 1394040-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110107

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110107
Availability details for
Quantity
  • Availability
  • Add to Cart