ENA371463746 Enamine

3-ethylpentanal

  • MDL number MFCD20432300
  • Cas Number : 39992-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110169
Availability details for
Quantity
  • Availability
  • Add to Cart