ENA371463749 Enamine

(4-amino-2-methoxyphenyl)urea

  • MDL number MFCD09047451
  • Cas Number : 926202-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108581
Availability details for
Quantity
  • Availability
  • Add to Cart