ENA371463750 Enamine

6-oxa-9-azaspiro[4.5]decane

  • MDL number MFCD09971189
  • Cas Number : 130643-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110174

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110174
Availability details for
Quantity
  • Availability
  • Add to Cart