ENA371463754 Enamine

1-amino-3-(2-bromophenoxy)propan-2-ol

  • MDL number MFCD00456166
  • Cas Number : 112169-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110178
Availability details for
Quantity
  • Availability
  • Add to Cart