ENA371463756 Enamine

4-methanesulfonylbutan-2-amine

  • MDL number MFCD16860302
  • Cas Number : 1247371-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108582
Availability details for
Quantity
  • Availability
  • Add to Cart