ENA371463758 Enamine

1-[2-chloro-4-(1H-1,2,4-triazol-1-yl)phenyl]ethan-1-one

  • MDL number MFCD20297096
  • Cas Number : 1343189-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110182
Availability details for
Quantity
  • Availability
  • Add to Cart