ENA371463767 Enamine

1-[2-chloro-4-(morpholin-4-yl)phenyl]ethan-1-ol

  • MDL number MFCD20297144
  • Cas Number : 1343761-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110193
Availability details for
Quantity
  • Availability
  • Add to Cart