ENA371463771 Enamine

1-(3-methylbutyl)-1H-pyrazol-3-amine

  • MDL number MFCD12149362
  • Cas Number : 1003012-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110224
Availability details for
Quantity
  • Availability
  • Add to Cart