ENA371463777 Enamine

2-(5,5-dimethyl-1,3-dioxan-2-yl)-2-methylpropan-1-ol

  • MDL number MFCD22378746
  • Cas Number : 7299-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110232
Availability details for
Quantity
  • Availability
  • Add to Cart