ENA371463779 Enamine

2-(6-methylpyridazin-3-yl)acetonitrile

  • MDL number MFCD18292025
  • Cas Number : 1394042-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108589
Availability details for
Quantity
  • Availability
  • Add to Cart