ENA371463780 Enamine

2-methyl-N-(1,2,3,4-tetrahydroquinolin-5-yl)propanamide

  • MDL number MFCD09805877
  • Cas Number : 1016526-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110238
Availability details for
Quantity
  • Availability
  • Add to Cart