ENA371463787 Enamine

2-(3-chloropropyl)-5-methyl-1,3,4-oxadiazole

  • MDL number MFCD12188457
  • Cas Number : 1153037-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110249
Availability details for
Quantity
  • Availability
  • Add to Cart