ENA371463789 Enamine

4-methyl-3-(1,3-oxazol-2-yl)aniline

  • MDL number MFCD14657181
  • Cas Number : 1150100-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110251
Availability details for
Quantity
  • Availability
  • Add to Cart