ENA371463797 Enamine

3-azaspiro[5.5]undecane-3-sulfonyl chloride

  • MDL number MFCD17253006
  • Cas Number : 1340329-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110261
Availability details for
Quantity
  • Availability
  • Add to Cart