ENA371463800 Enamine

2-propyl-2-(trifluoromethyl)pyrrolidine

  • MDL number MFCD21781768
  • Cas Number : 1394042-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110265
Availability details for
Quantity
  • Availability
  • Add to Cart