ENA371463806 Enamine

3-cyclohexyl-4-methyl-1,2-oxazol-5-amine

  • MDL number MFCD11209344
  • Cas Number : 909132-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-110309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110309
Availability details for
Quantity
  • Availability
  • Add to Cart