ENA371463807 Enamine

5-propyl-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD06803594
  • Cas Number : 31130-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110310
Availability details for
Quantity
  • Availability
  • Add to Cart