ENA371463810 Enamine

2-(3,3-dimethyl-2,5-dioxopyrrolidin-1-yl)acetic acid

  • MDL number MFCD21187182
  • Cas Number : 1337926-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110314
Availability details for
Quantity
  • Availability
  • Add to Cart