ENA371463812 Enamine

1-(3-chlorophenyl)cyclopentan-1-amine

  • MDL number MFCD09904114
  • Cas Number : 856563-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110316

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110316
Availability details for
Quantity
  • Availability
  • Add to Cart