ENA371463814 Enamine

3-(3-fluorophenyl)azetidine hydrochloride

  • MDL number MFCD08460713
  • Cas Number : 1203685-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110318

Safety Information

msds https://www.enaminestore.com/catalog/EN300-110318
Availability details for
Quantity
  • Availability
  • Add to Cart