ENA371463820 Enamine

2-(4-tert-butylcyclohexylidene)acetic acid

  • MDL number MFCD11124996
  • Cas Number : 13733-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110326
Availability details for
Quantity
  • Availability
  • Add to Cart