ENA371463822 Enamine

2-(oxane-4-sulfonyl)propanoic acid

  • MDL number MFCD17270406
  • Cas Number : 1394042-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-110328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110328
Availability details for
Quantity
  • Availability
  • Add to Cart