ENA371463824 Enamine

2-(5-methylfuran-2-yl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD14615235
  • Cas Number : 1250012-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-110332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-110332
Availability details for
Quantity
  • Availability
  • Add to Cart